siRNA Details
Gene Symbol: MAU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015329.3 | NP_056144.3 | 9, 10 | 1010 | ||
XM_005259837.3 | XP_005259894.2 | 9, 10 | 1002 | |||
XM_005259838.3 | XP_005259895.3 | 7, 8 | 1061 | |||
XM_006722711.3 | XP_006722774.2 | 9, 10 | 1002 | |||
XM_011527844.2 | XP_011526146.1 | 9, 10 | 1415 | |||
XM_017026539.1 | XP_016882028.1 | 7, 8 | 1060 | |||
GenBank mRNA | AB020699.1 | - | 9, 10 | 996 | ||
AK025896.1 | - | 7, 8 | 662 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
