siRNA Details
Gene Symbol: EPB41L5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184937.1 | NP_001171866.1 | 11 | 1008 | ||
NM_001184938.2 | NP_001171867.1 | 11 | 1298 | |||
NM_001184939.1 | NP_001171868.1 | 11 | 1008 | |||
NM_020909.3 | NP_065960.2 | 11 | 1008 | |||
NR_135920.1 | 11 | 1008 | ||||
XM_006712651.1 | XP_006712714.1 | 11 | 988 | |||
XM_006712652.1 | XP_006712715.1 | 11 | 1303 | |||
XM_006712653.1 | XP_006712716.1 | 11 | 1008 | |||
XM_011511529.2 | XP_011509831.1 | 11 | 988 | |||
XM_017004567.1 | XP_016860056.1 | 11 | 844 | |||
XM_017004568.1 | XP_016860057.1 | 11 | 1008 | |||
GenBank mRNA | AB046768.1 | - | 4 | 257 | ||
AK023019.1 | - | 11 | 959 | |||
AK290895.1 | - | 11 | 918 | |||
BC032822.2 | - | 11 | 1298 | 4827274 | ||
BC054508.1 | - | 11 | 943 | 5502663 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
