siRNA Details
Gene Symbol: ATF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256090.1 | NP_001243019.1 | 5 | 405 | ||
NM_001256091.1 | NP_001243020.1 | 4 | 330 | |||
NM_001256092.1 | NP_001243021.1 | 4 | 370 | |||
NM_001256093.1 | NP_001243022.1 | 4 | 370 | |||
NM_001256094.1 | NP_001243023.1 | 4 | 370 | |||
NM_001880.3 | NP_001871.2 | 4 | 370 | |||
NR_045768.1 | 5 | 405 | ||||
NR_045769.1 | 5 | 405 | ||||
NR_045770.1 | 5 | 405 | ||||
NR_045771.1 | 4 | 370 | ||||
NR_045772.1 | 5 | 405 | ||||
NR_045773.1 | 4 | 370 | ||||
NR_045774.1 | 4 | 370 | ||||
GenBank mRNA | AK128731.1 | - | 4 | 339 | ||
AY029364.1 | - | 2 | 71 | |||
BC026175.1 | - | 4 | 337 | |||
BC107698.1 | - | 4 | 337 | 6456836 | ||
BC130335.1 | - | 3 | 136 | 40146366 | ||
BC130337.1 | - | 3 | 136 | 40146368 | ||
BX648469.1 | - | 4 | 257 | |||
DA995464.1 | - | 4 | 370 | |||
DQ003036.1 | - | 5 | 368 | |||
DQ003037.1 | - | 4 | 333 | |||
DQ003038.1 | - | 4 | 333 | |||
DQ003039.1 | - | 4 | 333 | |||
DQ003040.1 | - | 5 | 368 | |||
DQ003041.1 | - | 5 | 368 | |||
DQ003042.1 | - | 5 | 368 | |||
DQ003043.1 | - | 5 | 368 | |||
DQ003044.1 | - | 5 | 368 | |||
DQ003045.1 | - | 4 | 333 | |||
DQ003046.1 | - | 4 | 333 | |||
DQ003047.1 | - | 4 | 333 | |||
DQ003048.1 | - | 4 | 333 | |||
DQ003049.1 | - | 4 | 293 | |||
X15875.1 | - | 2 | 97 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
