siRNA Details
Gene Symbol: ATF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256090.1 | NP_001243019.1 | 15 | 1715 | ||
NM_001256091.1 | NP_001243020.1 | 14 | 1640 | |||
NM_001256092.1 | NP_001243021.1 | 13 | 1583 | |||
NM_001256093.1 | NP_001243022.1 | 13 | 1615 | |||
NM_001880.3 | NP_001871.2 | 14 | 1680 | |||
NR_045768.1 | 17 | 1877 | ||||
NR_045769.1 | 17 | 1909 | ||||
NR_045770.1 | 16 | 1762 | ||||
NR_045771.1 | 15 | 1754 | ||||
NR_045772.1 | 15 | 1692 | ||||
NR_045773.1 | 14 | 1657 | ||||
NR_045774.1 | 13 | 1565 | ||||
GenBank mRNA | AK128731.1 | - | 14 | 1580 | ||
AY029364.1 | - | 4 | 298 | |||
BC107698.1 | - | 13 | 1550 | 6456836 | ||
BC130335.1 | - | 13 | 1446 | 40146366 | ||
BC130337.1 | - | 13 | 1446 | 40146368 | ||
BX648469.1 | - | 13 | 1470 | |||
DQ003036.1 | - | 17 | 1872 | |||
DQ003037.1 | - | 14 | 1643 | |||
DQ003038.1 | - | 6 | 560 | |||
DQ003039.1 | - | 15 | 1717 | |||
DQ003040.1 | - | 15 | 1655 | |||
DQ003041.1 | - | 15 | 1678 | |||
DQ003042.1 | - | 14 | 1581 | |||
DQ003043.1 | - | 17 | 1840 | |||
DQ003044.1 | - | 16 | 1725 | |||
DQ003045.1 | - | 13 | 1546 | |||
DQ003046.1 | - | 14 | 1620 | |||
DQ003047.1 | - | 13 | 1528 | |||
DQ003048.1 | - | 13 | 1578 | |||
DQ003049.1 | - | 14 | 1603 | |||
M31630.1 | - | 6 | 751 | |||
X15875.1 | - | 12 | 1407 |

More Information
Other Species Targeted

Gene Ontology Categories:
