siRNA Details
Gene Symbol: ATF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256090.1 | NP_001243019.1 | 15 | 2008 | ||
NM_001256091.1 | NP_001243020.1 | 14 | 1933 | |||
NM_001256092.1 | NP_001243021.1 | 13 | 1876 | |||
NM_001256093.1 | NP_001243022.1 | 13 | 1908 | |||
NM_001880.3 | NP_001871.2 | 14 | 1973 | |||
NR_045768.1 | 17 | 2170 | ||||
NR_045769.1 | 17 | 2202 | ||||
NR_045770.1 | 16 | 2055 | ||||
NR_045771.1 | 15 | 2047 | ||||
NR_045772.1 | 15 | 1985 | ||||
NR_045773.1 | 14 | 1950 | ||||
NR_045774.1 | 13 | 1858 | ||||
GenBank mRNA | AK128731.1 | - | 14 | 1875 | ||
BC107698.1 | - | 13 | 1843 | 6456836 | ||
BX648469.1 | - | 13 | 1762 | |||
M31630.1 | - | 6 | 1043 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
