siRNA Details
Gene Symbol: ADD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001119.4 | NP_001110.2 | 13 | 1824 | ||
NM_001286645.1 | NP_001273574.1 | 15 | 2592 | |||
NM_014189.3 | NP_054908.2 | 13 | 1917 | |||
NM_014190.3 | NP_054909.2 | 13 | 1824 | |||
NM_176801.2 | NP_789771.1 | 13 | 1917 | |||
XM_005247933.2 | XP_005247990.1 | 14 | 2135 | |||
XM_005247934.2 | XP_005247991.1 | 14 | 2099 | |||
XM_005247935.2 | XP_005247992.1 | 14 | 2042 | |||
XM_005247936.2 | XP_005247993.1 | 14 | 2135 | |||
XM_005247937.2 | XP_005247994.1 | 14 | 2042 | |||
XM_017007703.1 | XP_016863192.1 | 14 | 1860 | |||
XM_017007704.1 | XP_016863193.1 | 13 | 2006 | |||
XM_017007705.1 | XP_016863194.1 | 14 | 2006 | |||
XM_017007706.1 | XP_016863195.1 | 13 | 1913 | |||
XM_017007707.1 | XP_016863196.1 | 12 | 1671 | |||
XM_017007708.1 | XP_016863197.1 | 14 | 2042 | |||
XM_017007709.1 | XP_016863198.1 | 13 | 1913 | |||
XM_017007710.1 | XP_016863199.1 | 12 | 1671 | |||
GenBank mRNA | BC013393.2 | - | 13 | 1617 | 3532276 | |
BC042998.1 | - | 15 | 2597 | 5297337 | ||
CX783962.1 | - | 4 | 430 | |||
L07261.1 | - | 12 | 1731 | |||
S70312.1 | - | 3 | 141 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
