siRNA Details
Gene Symbol: ADD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001119.4 | NP_001110.2 | 14 | 1969 | ||
NM_001286645.1 | NP_001273574.1 | 16 | 2737 | |||
NM_014189.3 | NP_054908.2 | 14 | 2062 | |||
NM_014190.3 | NP_054909.2 | 14 | 1969 | |||
NM_176801.2 | NP_789771.1 | 14 | 2062 | |||
XM_005247933.2 | XP_005247990.1 | 15 | 2280 | |||
XM_005247934.2 | XP_005247991.1 | 15 | 2244 | |||
XM_005247935.2 | XP_005247992.1 | 15 | 2187 | |||
XM_005247936.2 | XP_005247993.1 | 15 | 2280 | |||
XM_005247937.2 | XP_005247994.1 | 15 | 2187 | |||
XM_017007703.1 | XP_016863192.1 | 15 | 2005 | |||
XM_017007704.1 | XP_016863193.1 | 14 | 2151 | |||
XM_017007705.1 | XP_016863194.1 | 15 | 2151 | |||
XM_017007706.1 | XP_016863195.1 | 14 | 2058 | |||
XM_017007707.1 | XP_016863196.1 | 13 | 1816 | |||
XM_017007708.1 | XP_016863197.1 | 15 | 2187 | |||
XM_017007709.1 | XP_016863198.1 | 14 | 2058 | |||
XM_017007710.1 | XP_016863199.1 | 13 | 1816 | |||
GenBank mRNA | BC013393.2 | - | 14 | 1762 | 3532276 | |
BC042998.1 | - | 16 | 2742 | 5297337 | ||
CX783962.1 | - | 5 | 575 | |||
L07261.1 | - | 13 | 1876 | |||
S70312.1 | - | 4 | 286 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
