siRNA Details
Gene Symbol: CNOT6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286790.1 | NP_001273719.1 | 11 | 1547 | ||
NM_144571.2 | NP_653172.2 | 11 | 1476 | |||
XM_011531806.2 | XP_011530108.1 | 12 | 1611 | |||
XM_011531807.2 | XP_011530109.1 | 12 | 1585 | |||
XM_011531808.2 | XP_011530110.1 | 11 | 1557 | |||
XM_011531809.1 | XP_011530111.1 | 11 | 1502 | |||
XM_011531810.1 | XP_011530112.1 | 11 | 2062 | |||
XM_017007956.1 | XP_016863445.1 | 11 | 1499 | |||
XM_017007957.1 | XP_016863446.1 | 11 | 1825 | |||
GenBank mRNA | AK027214.1 | - | 2 | 165 | ||
AK058188.1 | - | 11 | 1373 | |||
AK303417.1 | - | 11 | 1525 | |||
AL133112.1 | - | 5 | 774 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_178854, NM_001285511, NM_144910, NM_001285514, AK030112, and others.
Gene Ontology Categories:
