siRNA Details
Gene Symbol: PLEKHG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308147.1 | NP_001295076.1 | 15 | 1762 | ||
XM_011536624.2 | XP_011534926.1 | 17 | 2043 | |||
XM_011536625.2 | XP_011534927.1 | 17 | 1916 | |||
XM_011536627.2 | XP_011534929.1 | 17 | 2308 | |||
XM_011536628.2 | XP_011534930.1 | 17 | 2494 | |||
XM_011536629.2 | XP_011534931.1 | 16 | 1632 | |||
XM_017021154.1 | XP_016876643.1 | 16 | 1909 | |||
XM_017021155.1 | XP_016876644.1 | 17 | 1998 | |||
XM_017021156.1 | XP_016876645.1 | 17 | 2066 | |||
XM_017021157.1 | XP_016876646.1 | 15 | 1862 | |||
XM_017021158.1 | XP_016876647.1 | 15 | 1501 | |||
GenBank mRNA | AK074169.1 | - | 9 | 1533 | ||
BC004298.1 | - | 2 | 101 | |||
BC073907.1 | - | 2 | 50 | |||
BC129952.1 | - | 9 | 1254 | 40126555 | ||
BC129953.1 | - | 15 | 1573 | 40126558 | ||
BX248779.1 | - | 13 | 1566 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
