siRNA Details
Gene Symbol: MARVELD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017967.3 | NP_001017967.2 | 1 | 412 | ||
NM_052858.5 | NP_443090.4 | 1 | 412 | |||
XM_011523449.2 | XP_011521751.1 | 1 | 428 | |||
GenBank mRNA | AK056842.1 | - | 1 | 378 | ||
AK292185.1 | - | 1 | 378 | |||
BC005052.1 | - | 1 | 404 | |||
BC011893.2 | - | 1 | 379 | 3942185 | ||
BC013376.2 | - | 1 | 379 | 3946889 | ||
BM742281.1 | - | 1 | 398 | |||
BP320419.1 | - | 1 | 413 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
