siRNA Details
Gene Symbol: ZNF174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032292.2 | NP_001027463.1 | 1 | 1021 | ||
NM_003450.2 | NP_003441.1 | 1 | 1021 | |||
GenBank mRNA | AF542096.1 | - | 1 | 237 | ||
BC000876.1 | - | 1 | 948 | 3461658 | ||
BC001161.1 | - | 1 | 967 | 3533665 | ||
BT006923.1 | - | 1 | 206 | |||
U31248.1 | - | 1 | 791 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
