siRNA Details
Gene Symbol: SSH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282129.1 | NP_001269058.1 | 6 | 651 | ||
NM_001282130.1 | NP_001269059.1 | 5 | 570 | |||
NM_001282131.1 | NP_001269060.1 | 4 | 518 | |||
NM_033389.3 | NP_203747.2 | 5 | 382 | |||
XM_005258058.3 | XP_005258115.1 | 7 | 544 | |||
XM_005258059.2 | XP_005258116.1 | 4 | 518 | |||
XM_006722149.3 | XP_006722212.1 | 4 | 4149 | |||
XM_011525402.2 | XP_011523704.1 | 6 | 379 | |||
XM_011525404.2 | XP_011523706.1 | 4 | 1142 | |||
XM_011525405.2 | XP_011523707.1 | 4 | 1190 | |||
XM_011525406.2 | XP_011523708.1 | 3 | 469 | |||
XM_011525407.1 | XP_011523709.1 | 3 | 214 | |||
XM_017025251.1 | XP_016880740.1 | 6 | 590 | |||
XM_017025253.1 | XP_016880742.1 | 6 | 590 | |||
XM_017025254.1 | XP_016880743.1 | 6 | 590 | |||
XM_017025255.1 | XP_016880744.1 | 6 | 590 | |||
GenBank mRNA | AB072358.1 | - | 5 | 342 | ||
AB072359.1 | - | 5 | 553 | |||
AB099290.1 | - | 5 | 342 | |||
AK296413.1 | - | 6 | 516 | |||
AK308887.1 | - | 5 | 449 | |||
BC008941.2 | - | 5 | 494 | |||
BC011636.2 | - | 4 | 476 | |||
BC068223.1 | - | 5 | 494 | |||
DA861985.1 | - | 4 | 521 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
