siRNA Details
Gene Symbol: SSH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282129.1 | NP_001269058.1 | 7 | 754 | ||
NM_001282130.1 | NP_001269059.1 | 6 | 673 | |||
NM_001282131.1 | NP_001269060.1 | 5 | 621 | |||
NM_033389.3 | NP_203747.2 | 6 | 485 | |||
XM_005258058.3 | XP_005258115.1 | 8 | 647 | |||
XM_005258059.2 | XP_005258116.1 | 5 | 621 | |||
XM_006722149.3 | XP_006722212.1 | 5 | 4252 | |||
XM_011525402.2 | XP_011523704.1 | 7 | 482 | |||
XM_011525404.2 | XP_011523706.1 | 5 | 1245 | |||
XM_011525405.2 | XP_011523707.1 | 5 | 1293 | |||
XM_011525406.2 | XP_011523708.1 | 4 | 572 | |||
XM_011525407.1 | XP_011523709.1 | 4 | 317 | |||
XM_017025251.1 | XP_016880740.1 | 7 | 690 | |||
XM_017025252.1 | XP_016880741.1 | 5 | 551 | |||
XM_017025253.1 | XP_016880742.1 | 7 | 693 | |||
XM_017025254.1 | XP_016880743.1 | 7 | 693 | |||
XM_017025255.1 | XP_016880744.1 | 7 | 693 | |||
GenBank mRNA | AB072358.1 | - | 6 | 445 | ||
AB072359.1 | - | 6 | 656 | |||
AB099290.1 | - | 6 | 445 | |||
AK296413.1 | - | 7 | 619 | |||
AK308887.1 | - | 6 | 552 | |||
BC008941.2 | - | 6 | 597 | |||
BC011636.2 | - | 5 | 579 | |||
BC068223.1 | - | 6 | 597 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
