siRNA Details
Gene Symbol: TPGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033513.2 | NP_277048.2 | 1 | 36 | ||
GenBank mRNA | AK125401.1 | - | 1 | 33 | ||
AK126170.1 | - | 1 | 36 | |||
BM846833.1 | - | 1 | 36 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
