siRNA Details
Gene Symbol: DMRTC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040283.2 | NP_001035373.1 | 5 | 654 | ||
XM_011527214.1 | XP_011525516.1 | 5 | 666 | |||
XM_011527215.1 | XP_011525517.1 | 5 | 658 | |||
XM_011527216.2 | XP_011525518.1 | 5 | 713 | |||
XM_017027123.1 | XP_016882612.1 | 5 | 671 | |||
XM_017027124.1 | XP_016882613.1 | 5 | 673 | |||
XM_017027125.1 | XP_016882614.1 | 5 | 676 | |||
XM_017027126.1 | XP_016882615.1 | 5 | 766 | |||
XM_017027127.1 | XP_016882616.1 | 5 | 631 | |||
XM_017027128.1 | XP_016882617.1 | 5 | 836 | |||
XM_017027129.1 | XP_016882618.1 | 5 | 667 | |||
GenBank mRNA | AJ291669.1 | - | 4 | 470 | ||
AK057404.1 | - | 5 | 658 | |||
AK301822.1 | - | 5 | 657 | |||
BC029202.1 | - | 5 | 620 | 5170924 | ||
BC039266.1 | - | 5 | 622 | 5298983 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
