siRNA Details
Gene Symbol: DMRTC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040283.2 | NP_001035373.1 | 2 | 235 | ||
XM_011527214.1 | XP_011525516.1 | 2 | 207 | |||
XM_011527215.1 | XP_011525517.1 | 2 | 199 | |||
XM_011527216.2 | XP_011525518.1 | 2 | 254 | |||
XM_017027123.1 | XP_016882612.1 | 2 | 252 | |||
XM_017027124.1 | XP_016882613.1 | 2 | 254 | |||
XM_017027125.1 | XP_016882614.1 | 2 | 257 | |||
XM_017027126.1 | XP_016882615.1 | 2 | 347 | |||
XM_017027127.1 | XP_016882616.1 | 2 | 212 | |||
XM_017027128.1 | XP_016882617.1 | 2 | 417 | |||
XM_017027129.1 | XP_016882618.1 | 2 | 248 | |||
GenBank mRNA | AJ291669.1 | - | 1 | 51 | ||
AK057404.1 | - | 2 | 199 | |||
AK301822.1 | - | 2 | 238 | |||
BC029202.1 | - | 2 | 201 | 5170924 | ||
BC039266.1 | - | 2 | 203 | 5298983 | ||
DB026313.1 | - | 2 | 238 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
