siRNA Details
Gene Symbol: ZNF787 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002836.3 | NP_001002836.2 | 2, 3 | 285 | ||
XM_011526445.2 | XP_011524747.1 | 2, 3 | 1969 | |||
GenBank mRNA | AF000560.1 | - | 1, 2 | 82 | ||
BC012495.1 | - | 2, 3 | 189 | |||
BC077728.1 | - | 2, 3 | 192 | 6210543 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
