siRNA Details
Gene Symbol: PTH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178449.3 | NP_848544.1 | 1 | 133 | ||
GenBank mRNA | AY037555.1 | - | 1 | 133 | ||
AY048588.1 | - | 1 | 34 | |||
BC093995.1 | - | 1 | 72 | 7939840 | ||
BC093997.1 | - | 1 | 76 | 7939842 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
