siRNA Details
Gene Symbol: EVA1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304762.1 | NP_001291691.1 | 3 | 796 | ||
NM_018166.2 | NP_060636.1 | 3 | 868 | |||
NR_130899.1 | 2 | 699 | ||||
XM_017001627.1 | XP_016857116.1 | 2 | 990 | |||
GenBank mRNA | AK001509.1 | - | 3 | 868 | ||
BC006241.2 | - | 2 | 580 | 3951625 | ||
BC071697.1 | - | 2 | 787 | 4371667 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
