siRNA Details
Gene Symbol: FAHD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320848.1 | NP_001307777.1 | 3 | 339 | ||
NM_001320849.1 | NP_001307778.1 | 2 | 213 | |||
NM_199336.2 | NP_955368.1 | 2 | 213 | |||
XM_011510745.2 | XP_011509047.1 | 3 | 329 | |||
XM_011510746.2 | XP_011509048.1 | 3 | 448 | |||
XM_011510747.2 | XP_011509049.1 | 2 | 213 | |||
XM_011510748.2 | XP_011509050.1 | 3 | 329 | |||
XM_011510750.2 | XP_011509052.1 | 3 | 331 | |||
XM_017003470.1 | XP_016858959.1 | 3 | 326 | |||
XM_017003471.1 | XP_016858960.1 | 3 | 329 | |||
XM_017003472.1 | XP_016858961.1 | 2 | 205 | |||
XM_017003473.1 | XP_016858962.1 | 2 | 213 | |||
GenBank mRNA | AK295056.1 | - | 2 | 166 | ||
AL833869.1 | - | 1 | 13 | |||
BC064511.1 | - | 2 | 176 | 6644987 | ||
BI547630.1 | - | 2 | 218 | |||
HY033720.1 | - | 3 | 324 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
