siRNA Details
Gene Symbol: FAHD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320848.1 | NP_001307777.1 | 9 | 1415 | ||
NM_199336.2 | NP_955368.1 | 8 | 1289 | |||
GenBank mRNA | AL833869.1 | - | 7 | 1089 | ||
BC064511.1 | - | 8 | 1252 | 6644987 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
