siRNA Details
Gene Symbol: AGBL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178563.3 | NP_848658.3 | 4 | 534 | ||
XM_011516122.2 | XP_011514424.1 | 3 | 340 | |||
XM_017012136.1 | XP_016867625.1 | 4 | 1856 | |||
XM_017012137.1 | XP_016867626.1 | 4 | 2138 | |||
XM_017012138.1 | XP_016867627.1 | 3 | 340 | |||
XM_017012139.1 | XP_016867628.1 | 3 | 340 | |||
XM_017012140.1 | XP_016867629.1 | 3 | 340 | |||
XM_017012142.1 | XP_016867631.1 | 3 | 340 | |||
XM_017012143.1 | XP_016867632.1 | 3 | 340 | |||
GenBank mRNA | AK023045.1 | - | 4 | 472 | ||
AK302827.1 | - | 4 | 534 | |||
BC030651.2 | - | 4 | 509 | 5267615 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
