siRNA Details
Gene Symbol: RBMX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002139.3 | NP_002130.2 | 9 | 1625 | ||
NR_028476.1 | 8 | 1453 | ||||
NR_028477.1 | 9 | 1660 | ||||
GenBank mRNA | AK091520.1 | - | 8 | 1399 | ||
AK096015.1 | - | 2 | 1531 | |||
BC006550.2 | - | 9 | 1561 | 3010123 | ||
BC007435.2 | - | 9 | 1561 | 3010123 | ||
BX647131.1 | - | 7 | 2029 | |||
Z23064.1 | - | 8 | 1426 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
