siRNA Details
Gene Symbol: RBMX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002139.3 | NP_002130.2 | 9 | 1701 | ||
NR_028476.1 | 8 | 1529 | ||||
NR_028477.1 | 9 | 1736 | ||||
GenBank mRNA | AK091520.1 | - | 8 | 1475 | ||
AK096015.1 | - | 2 | 1607 | |||
BC006550.2 | - | 9 | 1637 | 3010123 | ||
BC007435.2 | - | 9 | 1637 | 3010123 | ||
BX647131.1 | - | 7 | 2105 | |||
Z23064.1 | - | 8 | 1502 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
