siRNA Details
Gene Symbol: KCNA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002235.3 | NP_002226.1 | 1 | 1425 | ||
XM_005253686.3 | XP_005253743.1 | 1 | 3724 | |||
XM_011520955.2 | XP_011519257.1 | 1 | 3724 | |||
XM_017019270.1 | XP_016874759.1 | 1 | 3724 | |||
XM_017019271.1 | XP_016874760.1 | 1 | 3724 | |||
XM_017019272.1 | XP_016874761.1 | 1 | 3724 | |||
GenBank mRNA | BC069355.1 | - | 1 | 656 | 7262373 | |
X17622.1 | - | 1 | 1421 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
