siRNA Details
Gene Symbol: UTRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007124.2 | NP_009055.2 | 8 | 842 | ||
XM_005267127.4 | XP_005267184.1 | 8 | 1177 | |||
XM_005267130.2 | XP_005267187.1 | 9 | 1060 | |||
XM_005267133.2 | XP_005267190.1 | 8 | 1860 | |||
XM_011536101.2 | XP_011534403.1 | 10 | 1521 | |||
XM_011536102.2 | XP_011534404.1 | 10 | 1062 | |||
XM_011536106.2 | XP_011534408.1 | 9 | 1709 | |||
XM_011536107.2 | XP_011534409.1 | 9 | 1085 | |||
XM_017011243.1 | XP_016866732.1 | 10 | 1062 | |||
XM_017011244.1 | XP_016866733.1 | 10 | 1062 | |||
XM_017011245.1 | XP_016866734.1 | 10 | 1062 | |||
GenBank mRNA | AK292506.1 | - | 9 | 1060 | ||
BX472896.1 | - | 3 | 238 | |||
X69086.1 | - | 8 | 750 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
