siRNA Details
Gene Symbol: RANBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005493.2 | NP_005484.2 | 4 | 838 | ||
XM_006714945.2 | XP_006715008.1 | 4 | 318 | |||
XM_011514205.2 | XP_011512507.1 | 4 | 856 | |||
XM_017010149.1 | XP_016865638.1 | 4 | 226 | |||
GenBank mRNA | AB008515.1 | - | 4 | 527 | ||
AB055311.1 | - | 4 | 838 | |||
AF064606.1 | - | 4 | 333 | |||
AF306510.1 | - | 4 | 376 | |||
AK313334.1 | - | 3 | 229 | |||
BC019886.1 | - | 4 | 292 | |||
BC052781.1 | - | 4 | 999 | |||
BC063849.1 | - | 3 | 117 | 6186252 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
