siRNA Details
Gene Symbol: RANBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005493.2 | NP_005484.2 | 9, 10 | 1592 | ||
XM_006714945.2 | XP_006715008.1 | 9, 10 | 1072 | |||
XM_017010149.1 | XP_016865638.1 | 9, 10 | 980 | |||
GenBank mRNA | AB008515.1 | - | 9, 10 | 1281 | ||
AB055311.1 | - | 9, 10 | 1592 | |||
AF064606.1 | - | 7, 8 | 604 | |||
AF306510.1 | - | 9, 10 | 1130 | |||
AK313334.1 | - | 8, 9 | 983 | |||
BC019886.1 | - | 9, 10 | 1046 | |||
BC052781.1 | - | 9, 10 | 1753 | |||
BC063849.1 | - | 7, 8 | 848 | 6186252 | ||
KU921435.1 | - | 4, 5 | 606 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
