siRNA Details
Gene Symbol: ALX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006492.2 | NP_006483.2 | 4 | 1045 | ||
GenBank mRNA | AF008203.1 | - | 4 | 957 | ||
BC112007.1 | - | 4 | 1045 | 8327475 | ||
BC113428.1 | - | 4 | 1045 | 8322480 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
