siRNA Details
Gene Symbol: ANKRD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152326.2 | NP_689539.1 | 4 | 1532 | ||
XM_005267310.2 | XP_005267367.1 | 3 | 1669 | |||
XM_005267311.3 | XP_005267368.1 | 3 | 1301 | |||
GenBank mRNA | AK291868.1 | - | 3 | 1198 | ||
BC015422.1 | - | 2 | 1331 | 4397794 | ||
BC027479.1 | - | 2 | 1049 | 5114925 | ||
BX161460.1 | - | 4 | 1532 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
