siRNA Details
Gene Symbol: ANKRD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005267310.2 | XP_005267367.1 | 3 | 1767 | ||
XM_005267311.3 | XP_005267368.1 | 3 | 1399 | |||
GenBank mRNA | AK291868.1 | - | 3 | 1296 | ||
BC015422.1 | - | 2 | 1429 | 4397794 | ||
BC027479.1 | - | 2 | 1147 | 5114925 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
