siRNA Details
Gene Symbol: SST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048.3 | NP_001039.1 | 2 | 367 | ||
GenBank mRNA | AK312177.1 | - | 2 | 346 | ||
BC032625.1 | - | 2 | 272 | 5240781 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
