siRNA Details
Gene Symbol: CELF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172648.3 | NP_001166119.1 | 5 | 1008 | ||
NM_001172649.3 | NP_001166120.1 | 5 | 1255 | |||
NM_001291106.1 | NP_001278035.1 | 5 | 1255 | |||
NM_001291107.1 | NP_001278036.1 | 5 | 1252 | |||
NM_007185.6 | NP_009116.3 | 5 | 1255 | |||
XM_006711139.2 | XP_006711202.1 | 6 | 483 | |||
XM_006711141.2 | XP_006711204.1 | 6 | 480 | |||
XM_011509111.2 | XP_011507413.1 | 5 | 1103 | |||
XM_011509112.2 | XP_011507414.1 | 4 | 346 | |||
XM_011509114.2 | XP_011507416.1 | 4 | 314 | |||
XM_017000145.1 | XP_016855634.1 | 4 | 321 | |||
XM_017000146.1 | XP_016855635.1 | 4 | 318 | |||
GenBank mRNA | AB209264.1 | - | 5 | 1008 | ||
AF329264.1 | - | 5 | 455 | |||
AL519587.3 | - | 6 | 487 | |||
AY165003.1 | - | 5 | 1245 | |||
BC004145.2 | - | 6 | 430 | |||
BC041088.1 | - | 5 | 487 | |||
BC068008.1 | - | 4 | 282 | |||
BC104758.1 | - | 5 | 518 | 8143761 | ||
BC143226.1 | - | 5 | 515 | 9051727 | ||
U80746.1 | - | 2 | 134 | |||
U80759.1 | - | 2 | 141 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
