siRNA Details
Gene Symbol: SHF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301168.1 | NP_001288097.1 | 4 | 886 | ||
NM_001301169.1 | NP_001288098.1 | 4 | 886 | |||
NM_001301171.1 | NP_001288100.1 | 4 | 473 | |||
NM_138356.2 | NP_612365.2 | 6 | 1311 | |||
XM_006720742.3 | XP_006720805.2 | 5 | 1026 | |||
XM_011522174.2 | XP_011520476.1 | 5 | 1026 | |||
XM_011522175.2 | XP_011520477.1 | 5 | 1026 | |||
XM_017022711.1 | XP_016878200.1 | 5 | 1137 | |||
GenBank mRNA | AK094391.1 | - | 5 | 755 | ||
AK096267.1 | - | 6 | 1311 | |||
AK123658.1 | - | 1 | 374 | |||
AK301625.1 | - | 4 | 473 | |||
AK303749.1 | - | 5 | 661 | |||
BC007586.1 | - | 3 | 248 | |||
BC101601.1 | - | 4 | 467 | 8069107 | ||
BC101603.1 | - | 4 | 466 | 8069109 | ||
BC143601.1 | - | 4 | 467 | |||
BC171730.1 | - | 3 | 326 | |||
BX332806.2 | - | 4 | 656 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
