siRNA Details
Gene Symbol: KRT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256282.1 | NP_001243211.1 | 6 | 908 | ||
NM_001256293.1 | NP_001243222.1 | 6 | 1002 | |||
NM_002273.3 | NP_002264.1 | 5 | 907 | |||
NR_045962.1 | 6 | 1261 | ||||
GenBank mRNA | AK222941.1 | - | 5 | 880 | ||
AK290938.1 | - | 6 | 960 | |||
AK310257.1 | - | 6 | 908 | |||
AK315826.1 | - | 6 | 1237 | |||
BC000654.2 | - | 5 | 868 | 3349233 | ||
BC008200.1 | - | 5 | 522 | 3445666 | ||
BC011373.1 | - | 4 | 450 | |||
BC063513.1 | - | 5 | 872 | 3879823 | ||
BC073760.1 | - | 5 | 854 | 2963092 | ||
BC075839.1 | - | 5 | 852 | 6285632 | ||
BX417476.2 | - | 5 | 895 | |||
M34225.1 | - | 5 | 838 | |||
U76549.1 | - | 5 | 820 | |||
X12882.1 | - | 2 | 192 | |||
X74929.1 | - | 5 | 863 | |||
X98614.1 | - | 2 | 192 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
