siRNA Details
Gene Symbol: KRT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256282.1 | NP_001243211.1 | 9 | 1621 | ||
NM_001256293.1 | NP_001243222.1 | 9 | 1715 | |||
NM_002273.3 | NP_002264.1 | 8 | 1620 | |||
NR_045962.1 | 9 | 1974 | ||||
GenBank mRNA | AK222941.1 | - | 8 | 1593 | ||
AK290938.1 | - | 9 | 1673 | |||
AK310257.1 | - | 9 | 1621 | |||
AK315826.1 | - | 9 | 1950 | |||
BC000654.2 | - | 8 | 1581 | 3349233 | ||
BC008200.1 | - | 8 | 1235 | 3445666 | ||
BC011373.1 | - | 7 | 1163 | |||
BC063513.1 | - | 8 | 1585 | 3879823 | ||
BC073760.1 | - | 8 | 1567 | 2963092 | ||
BC075839.1 | - | 8 | 1565 | 6285632 | ||
CX166336.1 | - | 4 | 595 | |||
M34225.1 | - | 8 | 1551 | |||
M77025.1 | - | 8 | 1222 | |||
X12882.1 | - | 5 | 905 | |||
X74929.1 | - | 8 | 1576 | |||
X98614.1 | - | 5 | 905 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
