siRNA Details
Gene Symbol: FLYWCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142499.1 | NP_001135971.1 | 4 | 846 | ||
NM_001142500.1 | NP_001135972.1 | 4 | 936 | |||
NM_138439.2 | NP_612448.1 | 4 | 940 | |||
XM_005255078.4 | XP_005255135.1 | 3 | 1143 | |||
GenBank mRNA | AL542248.3 | - | 4 | 830 | ||
BC014089.2 | - | 4 | 815 | 4330613 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
