siRNA Details
Gene Symbol: ZYG11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024646.2 | NP_078922.1 | 3 | 850 | ||
XM_006710898.3 | XP_006710961.1 | 3 | 737 | |||
XM_017002336.1 | XP_016857825.1 | 3 | 855 | |||
GenBank mRNA | AB051517.1 | - | 3 | 800 | ||
AK095643.1 | - | 2 | 490 | |||
AK296460.1 | - | 3 | 855 | |||
BX647643.1 | - | 1 | 23 | |||
DQ646393.1 | - | 3 | 869 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
