siRNA Details
Gene Symbol: ZYG11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024646.2 | NP_078922.1 | 14 | 2356 | ||
XM_006710898.3 | XP_006710961.1 | 14 | 2243 | |||
GenBank mRNA | AB051517.1 | - | 14 | 2306 | ||
AK023518.1 | - | 8 | 813 | |||
AK056645.1 | - | 12 | 1300 | |||
AK095643.1 | - | 13 | 1996 | |||
AK296460.1 | - | 12 | 2151 | |||
BC029832.1 | - | 12 | 1370 | |||
BX647643.1 | - | 12 | 1529 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
