siRNA Details
Gene Symbol: LYSMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136543.1 | NP_001130015.1 | 3 | 1277 | ||
NM_212551.4 | NP_997716.1 | 3 | 1891 | |||
GenBank mRNA | AK298814.1 | - | 3 | 1279 | ||
AL122088.1 | - | 2 | 874 | |||
AY037156.1 | - | 3 | 1352 | |||
BC031649.1 | - | 3 | 1495 | 5172124 | ||
BX647911.1 | - | 3 | 1857 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
