siRNA Details
Gene Symbol: LYSMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136543.1 | NP_001130015.1 | 3 | 1664 | ||
NM_212551.4 | NP_997716.1 | 3 | 2278 | |||
GenBank mRNA | AL122088.1 | - | 2 | 1268 | ||
AY037156.1 | - | 3 | 1738 | |||
BC031649.1 | - | 3 | 1891 | 5172124 | ||
BX647911.1 | - | 3 | 2252 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
