siRNA Details
Gene Symbol: CAMK2N2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033259.2 | NP_150284.1 | 1 | 336 | ||
GenBank mRNA | AY037149.1 | - | 1 | 336 | ||
BC105077.1 | - | 1 | 248 | 8144080 | ||
BC105079.1 | - | 1 | 248 | 8144082 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
