siRNA Details
Gene Symbol: CLEC2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004419.4 | NP_001004419.1 | 6 | 2231 | ||
NM_001197317.2 | NP_001184246.1 | 4 | 2038 | |||
NM_001197318.2 | NP_001184247.1 | 4 | 2045 | |||
NM_001197319.2 | NP_001184248.1 | 3 | 1934 | |||
NM_013269.5 | NP_037401.1 | 5 | 2149 | |||
NR_036693.2 | 4 | 2120 |
Gene Symbol: NPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002520.6 | NP_002511.1 | 11 | 1320 | ||
NM_199185.3 | NP_954654.1 | 10 | 1233 | |||
XM_011534564.1 | XP_011532866.1 | 10 | 1140 | |||
GenBank mRNA | AY347529.1 | - | 12 | 1210 | ||
BC002398.2 | - | 11 | 1162 | 2821577 | ||
BC003670.1 | - | 1 | 877 | |||
BC008495.1 | - | 11 | 1172 | 4276604 | ||
BC009623.2 | - | 10 | 1113 | 3877633 | ||
BC012566.1 | - | 11 | 1174 | 4097025 | ||
BC014349.1 | - | 11 | 1171 | 3511037 | ||
BC016716.1 | - | 11 | 1174 | 4068167 | ||
BC016768.1 | - | 11 | 1173 | 4081364 | ||
BC016824.1 | - | 11 | 1174 | 4106286 | ||
BC020467.1 | - | 11 | 1160 | |||
BC021668.1 | - | 11 | 1175 | 3930893 | ||
BC021983.1 | - | 11 | 1173 | 4106023 | ||
BC050628.1 | - | 11 | 1161 | 5575414 | ||
DQ303464.1 | - | 11 | 1175 | |||
M23613.1 | - | 11 | 1168 | |||
M26697.1 | - | 11 | 1165 | |||
M28699.1 | - | 11 | 1162 | |||
X16934.1 | - | 11 | 1035 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
