siRNA Details
Gene Symbol: NPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002520.6 | NP_002511.1 | 11 | 1370 | ||
NM_199185.3 | NP_954654.1 | 10 | 1283 | |||
XM_011534564.1 | XP_011532866.1 | 10 | 1190 | |||
GenBank mRNA | AY347529.1 | - | 12 | 1260 | ||
BC002398.2 | - | 11 | 1212 | 2821577 | ||
BC003670.1 | - | 1 | 927 | |||
BC008495.1 | - | 11 | 1222 | 4276604 | ||
BC009623.2 | - | 10 | 1163 | 3877633 | ||
BC012566.1 | - | 11 | 1224 | 4097025 | ||
BC014349.1 | - | 11 | 1221 | 3511037 | ||
BC016716.1 | - | 11 | 1224 | 4068167 | ||
BC016768.1 | - | 11 | 1223 | 4081364 | ||
BC016824.1 | - | 11 | 1224 | 4106286 | ||
BC020467.1 | - | 11 | 1210 | |||
BC021668.1 | - | 11 | 1225 | 3930893 | ||
BC021983.1 | - | 11 | 1223 | 4106023 | ||
BC050628.1 | - | 11 | 1211 | 5575414 | ||
DQ303464.1 | - | 11 | 1225 | |||
M23613.1 | - | 11 | 1218 | |||
M26697.1 | - | 11 | 1215 | |||
M28699.1 | - | 11 | 1212 | |||
X16934.1 | - | 11 | 1085 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
