siRNA Details
Gene Symbol: INTS4P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027392.2 | 9, 10 | 1316 | |||
GenBank mRNA | BC111554.1 | - | 9, 10 | 1309 | 40024359 |
Gene Symbol: INTS4P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057766.1 | - | 9, 10 | 1309 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
