siRNA Details
Gene Symbol: PGAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000290.3 | NP_000281.2 | 1 | 275 | ||
GenBank mRNA | BC001904.1 | - | 1 | 283 | 3535570 | |
BC073741.1 | - | 1 | 271 | 2823398 | ||
CT002473.1 | - | 1 | 275 | |||
M18172.1 | - | 1 | 252 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
