siRNA Details
Gene Symbol: CNOT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265612.1 | NP_001252541.1 | 45 | 6860 | ||
NM_016284.4 | NP_057368.3 | 45 | 6875 | |||
NR_049763.1 | 46 | 7043 | ||||
GenBank mRNA | AB023224.2 | - | 32 | 4920 | ||
AF110778.1 | - | 5 | 652 | |||
AK056927.1 | - | 22 | 3205 | |||
AK075125.1 | - | 15 | 2174 | |||
AK092885.1 | - | 5 | 1419 | |||
AK301092.1 | - | 5 | 639 | |||
AK302495.1 | - | 22 | 3245 | |||
AL117492.1 | - | 17 | 2714 | |||
AL832769.1 | - | 45 | 6824 | |||
AL833549.1 | - | 45 | 6834 | |||
BC000779.2 | - | 7 | 1033 | |||
BC024317.1 | - | 28 | 4274 | |||
BC027969.2 | - | 4 | 489 | |||
BX537840.1 | - | 46 | 6989 | |||
CR749237.1 | - | 38 | 5815 | |||
EF553522.1 | - | 45 | 6809 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
