siRNA Details
Gene Symbol: VSTM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031746.4 | NP_001026916.2 | 2 | 387 | ||
NM_144984.3 | NP_659421.1 | 2 | 387 | |||
XM_017015825.1 | XP_016871314.1 | 2 | 427 | |||
XM_017015826.1 | XP_016871315.1 | 2 | 427 | |||
XM_017015827.1 | XP_016871316.1 | 2 | 367 | |||
XM_017015828.1 | XP_016871317.1 | 2 | 807 | |||
GenBank mRNA | AK056299.1 | - | 2 | 364 | ||
AK297931.1 | - | 2 | 362 | |||
BC041414.1 | - | 2 | 369 | 5285468 | ||
DA920289.1 | - | 2 | 364 | |||
HY112498.1 | - | 2 | 388 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
