siRNA Details
Gene Symbol: POTED | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174981.3 | NP_778146.2 | 1 | 82 | ||
XM_006723997.3 | XP_006724060.1 | 1 | 178 | |||
XM_011529550.2 | XP_011527852.1 | 1 | 178 |
Gene Symbol: LOC100288966 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257362.1 | NP_001244291.1 | 1 | 82 | ||
XM_006709930.3 | XP_006709993.1 | 1 | 178 | |||
XM_011545655.2 | XP_011543957.1 | 1 | 178 | |||
XM_011545656.2 | XP_011543958.1 | 1 | 178 | |||
GenBank mRNA | AY172978.1 | - | 1 | 82 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
