siRNA Details
Gene Symbol: GNG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033258.1 | NP_150283.1 | 1 | 71 | ||
XM_011527524.2 | XP_011525826.1 | 2 | 631 | |||
XM_017027503.1 | XP_016882992.1 | 2 | 210 | |||
XM_017027504.1 | XP_016882993.1 | 2 | 2865 | |||
XM_017027505.1 | XP_016882994.1 | 2 | 164 | |||
GenBank mRNA | AF493875.1 | - | 1 | 71 | ||
BC095514.1 | - | 1 | 74 | 6519176 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
